Thiophene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 110-02-1
Molecular Formula: C4H4S
Formula Weight: 84.14
Flash Point: -9°(16°F)
Refractive Index: 1.5280
Odor: Stench
Melting Point: -38°
Boiling Point: 83-85°
Beilstein Reference: 103222
Merck Reference: 14,9353
UN No.: 2414
MDL No.: MFCD00005413
Formula:
C₄H₄S Boiling Pt: 83…85 °C Melting Pt: –38 °C Density: 1.066 Flash Pt: –9 °C (16 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005413 CAS Number: 110-02-1 EINECS: 203-729-4 UN: 2414 ADR: 3,II Merck Index: 14,9353 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...