Glycine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
A13816-0C
A13816-0E
A13816-36
AAAA13816-0CEA
513
USD
AAAA13816-0C
AAAA13816-0E
AAAA13816-36
Glycine 99%
Glycine
Purity: 99%
CAS No.: 56-40-6
Molecular Formula: NH2CH2CO2H
Formula Weight: 75.07
Odor: Odorless
pH: 5,97
Melting Point: ca 245° dec.
Beilstein Reference: 635782
Merck Reference: 14,4491
MDL No.: MFCD00008131
Formula:
NH₂CH₂COOH Boiling Pt: 181 °C (1013 hPa) Melting Pt: ∼ 245 °C (dec.) Density: 1.595 Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 14,4491 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...