Glycine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

A13816-0C A13816-0E A13816-36
AAAA13816-0CEA 513 USD
AAAA13816-0C AAAA13816-0E AAAA13816-36
Glycine 99%
Glycine

Purity: 99% CAS No.: 56-40-6 Molecular Formula: NH2CH2CO2H Formula Weight: 75.07 Odor: Odorless pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131

Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: ∼ 245 °C (dec.)
Density: 1.595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR