Ammonium heptamolybdate tetrahydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A13766-30 A13766-0B A13766-18
AAAA13766-30EA 148.74 USD
AAAA13766-30 AAAA13766-0B AAAA13766-18
Ammonium heptamolybdate tetrahydrate 99%
Ammonium heptamolybdate tetrahydrate

Purity: 99% CAS No.: 12054-85-2 Molecular Formula: (NH4)6Mo7O24.4H2O Formula Weight: 1235.86 (1163.79anhy) Melting Point: ca 190° dec. Merck Reference: 14,533 MDL No.: MFCD00167059

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Melting Pt: ∼ 190 °C (dec.)
Density: 2.498
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 14,533

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR