Ammonium heptamolybdate tetrahydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13766-30
A13766-0B
A13766-18
AAAA13766-30EA
148.74
USD
AAAA13766-30
AAAA13766-0B
AAAA13766-18
Ammonium heptamolybdate tetrahydrate 99%
Ammonium heptamolybdate tetrahydrate
Purity: 99%
CAS No.: 12054-85-2
Molecular Formula: (NH4)6Mo7O24.4H2O
Formula Weight: 1235.86 (1163.79anhy)
Melting Point: ca 190° dec.
Merck Reference: 14,533
MDL No.: MFCD00167059
Formula:
(NH₄)₆Mo₇O₂₄·4H₂O Melting Pt: ∼ 190 °C (dec.) Density: 2.498 Storage Temperature: Ambient |
MDL Number:
MFCD00167059 CAS Number: 12054-85-2 EINECS: 234-722-4 Merck Index: 14,533 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...