Triiodomethane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Iodoform
A13664-0B
A13664-30
A13664-18
AAAA13664-0BEA
529
USD
AAAA13664-0B
AAAA13664-30
AAAA13664-18
Triiodomethane 99%
Triiodomethane
Purity: 99%
CAS No.: 75-47-8
Molecular Formula: CHI3
Formula Weight: 393.73
Flash Point: None
Odor: Unpleasant
Beilstein Reference: 1697010
Merck Reference: 14,5033
MDL No.: MFCD00001069
Formula:
CHI₃ Boiling Pt: 218 °C (1013 hPa) Melting Pt: 118…123 °C Density: 4.008 Storage Temperature: Ambient |
MDL Number:
MFCD00001069 CAS Number: 75-47-8 EINECS: 200-874-5 Merck Index: 14,5033 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...