2-Bromopyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13241-36
A13241-30
A13241-14
A13241-22
AAA13241-36EA
386
USD
AAA13241-36
AAAA13241-30
AAA13241-14
AAA13241-22
2-Bromopyridine 99%
2-Bromopyridine
Purity: 99%
CAS No.: 109-04-6
Molecular Formula: C5H4BrN
Formula Weight: 158.00
Flash Point: 97°(207°F)
Refractive Index: 1.5720
Odor: Characteristic
Boiling Point: 192-194°
Beilstein Reference: 105789
UN No.: 2810
MDL No.: MFCD00006219
Formula:
C₅H₄BrN Boiling Pt: 192…194 °C Melting Pt: –40.1 °C Density: 1.625 Flash Pt: 97 °C (207 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006219 CAS Number: 109-04-6 EINECS: 203-641-6 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...