L-Phenylalanine 99%
Supplier: Thermo Scientific Chemicals
Purity: 99%
CAS No.: 63-91-2
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Physical Form: Cryst powder
Appearance: White
Melting Point: ca 275° dec.
Beilstein Reference: 1910408
Merck Reference: 14,7271
Optical Rotation: -34° (c=2 in water)
MDL No.: MFCD00064227
Formula:
C₉H₁₁NO₂ Boiling Pt: 313 °C (1013 hPa) Melting Pt: ∼ 275 °C (dec.) Density: 0.754 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064227 CAS Number: 63-91-2 EINECS: 200-568-1 Merck Index: 14,7271 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...