1,10-Phenanthroline, (max. 1,5% H₂O) 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Phenanthroline

A13163-06 A13163-14 A13163-22
AAAA13163-06EA 42.9 USD
AAAA13163-06 AAAA13163-14 AAAA13163-22
1,10-Phenanthroline, (max. 1,5% H₂O) 99%
1,10-Phenanthroline

1,10-Phenanthroline, anhydrous is used as a chelating agent, used as bidendate nitrogen ligands in Heck type reactions as an alternative to phospine ligands. It is widely used as an indicator for alkyllithium reagents and an inhibitor of metallopeptidase enzymes. It is also used in the analysis formulations in end-user industries like food, beverage and pharmaceuticals.


Purity: 99% CAS No.: 66-71-7 Molecular Formula: C12H8N2 Formula Weight: 180.21 Odor: Odorless Melting Point: 116-119° Boiling Point: 330° Beilstein Reference: 126461 Merck Reference: 14,7214 UN No.: 2811 MDL No.: MFCD00011678

Formula: C₁₂H₈N₂
Boiling Pt: >330 °C
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00011678
CAS Number: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,7214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR