3-Hydroxy-p-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

A12866-22 A12866-06 A12866-14
AAA12866-22EA 182 USD
AAA12866-22 AAAA12866-06 AAA12866-14
3-Hydroxy-p-anisaldehyde 98%
3-Hydroxy-p-anisaldehyde

Purity: 98% CAS No.: 621-59-0 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 110°(230°F) Melting Point: 112-116° Boiling Point: 179°/15mm Beilstein Reference: 1073021 MDL No.: MFCD00003369

Formula: C₈H₈O₃
Boiling Pt: 179 °C/15 mm
Melting Pt: 112…116 °C
Density: 1,20
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR