Ammonium iodide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12498-22 A12498-0E A12498-36
AAAA12498-22EA 64.3 USD
AAAA12498-22 AAAA12498-0E AAAA12498-36
Ammonium iodide 98+%
Ammonium iodide

Purity: 98+% CAS No.: 12027-06-4 Molecular Formula: NH4I Formula Weight: 144.94 Refractive Index: 1.7031 Odor: Odorless Merck Reference: 14,529 MDL No.: MFCD00011424

Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551 °C (subl.)
Density: 2.514
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 14,529

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR