Butyramide 98%

Supplier: Thermo Scientific Chemicals
Warning

A12494-18 A12494-22 A12494-36
AAAA12494-18EA 48 USD
AAAA12494-18 AAA12494-0E AAA12494-22 AAA12494-36
Butyramide 98%
Butyramide

Purity: 98% CAS No.: 541-35-5 Molecular Formula: CH3CH2CH2CONH2 Formula Weight: 87.12 Melting Point: 113-116° Boiling Point: 216° Merck Reference: 14,1592 MDL No.: MFCD00041894

Formula: C₄H₉NO
Boiling Pt: 216 °C
Melting Pt: 113…116 °C
Density: 1.032
Storage Temperature: Ambient
MDL Number: MFCD00041894
CAS Number: 541-35-5
EINECS: 208-776-4
Merck Index: 14,1592

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR