5-Amino-3-methylisoxazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

A12446-14 A12446-03 A12446-06
AAA12446-14EA 419 USD
AAA12446-14 AAAA12446-03 AAA12446-06
5-Amino-3-methylisoxazole 98%
5-Amino-3-methylisoxazole

Purity: 98+% CAS No.: 14678-02-5 Molecular Formula: C4H6N2O Formula Weight: 98.11 Melting Point: 81-85° Beilstein Reference: 107906

Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
Melting Pt: 84
Storage Temperature: Ambient
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR