Iodic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A11925-0B A11925-18 A11925-30
AAAA11925-0BEA 844.27 USD
AAAA11925-0B AAAA11925-18 AAAA11925-30
Iodic acid 99%
Iodic acid

Purity: 99% CAS No.: 7782-68-5 Molecular Formula: HIO3 Formula Weight: 175.91 Odor: Odorless Melting Point: 110° dec. UN No.: 3085 MDL No.: MFCD00011348

Formula: HIO₃
MW: 175.91 g/mol
Melting Pt: 110 (dec.)
Density: 4.629
Storage Temperature: Ambient
MDL Number: MFCD00011348
CAS Number: 7782-68-5
EINECS: 231-962-1
UN: 3085
ADR: 5.1 III
Merck Index: 14,5012

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR