Barium sulfate 99%, precipitated

Supplier: Thermo Scientific Chemicals

A11768-0E A11768-0C A11768-36
AAAA11768-0EEA 126 USD
AAAA11768-0E AAAA11768-0C AAAA11768-36
Barium sulfate 99%, precipitated
Barium sulphate

Purity: 99% CAS No.: 7727-43-7 Molecular Formula: BaSO4 Formula Weight: 233.40 Odor: Odorless Melting Point: 1580° dec. MDL No.: MFCD00003455

Formula: BaSO₄
MW: 233.39 g/mol
Boiling Pt: 1600
Melting Pt: 1345 (dec.)
Density: 4.5
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 14,994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR