3-Fluoropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11693-14
AAAA11693-14EA
253
USD
AAAA11693-14
3-Fluoropyridine 98%
3-Fluoropyridine
MDL: MFCD00006374
Beilstein Registry No.: 105702
Purity: 98%
CAS No.: 372-47-4
Molecular Formula: C5H4FN
Formula Weight: 97.09
Flash Point: 13°(55°F)
Refractive Index: 1.4720
Boiling Point: 106-108°
Beilstein Reference: 105702
UN No.: 1993
MDL No.: MFCD00006374
Formula:
C₅H₄FN Boiling Pt: 106…108 °C Density: 1.13 Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006374 CAS Number: 372-47-4 EINECS: 206-755-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...