Tin(II) oxalate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tin (II) ethanedioate, Oxalic acid tin (II) salt
A11689-36
A11689-0E
A11689-22
AAAA11689-36EA
85.5
USD
AAAA11689-36
AAAA11689-0E
AAAA11689-22
Tin(II) oxalate 98%
Tin (II) oxalate
Purity: 98%
CAS No.: 814-94-8
Molecular Formula: C2O4Sn
Formula Weight: 206.72
Melting Point: 280° dec.
Beilstein Reference: 3708588
Merck Reference: 14,8786
UN No.: 3288
MDL No.: MFCD00040678
Formula:
C₂O₄Sn MW: 206.73 g/mol Melting Pt: 280 °C (dec.) Density: 3.56 Storage Temperature: Ambient |
MDL Number:
MFCD00040678 CAS Number: 814-94-8 EINECS: 212-414-0 UN: 3288 ADR: 6.1 III Merck Index: 14,8786 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...