Lauric acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11672-0C
A11672-36
A11672-0E
AAAA11672-0CEA
232.42
USD
AAAA11672-0C
AAAA11672-36
AAAA11672-0E
Lauric acid 98%
Lauric acid
Purity: 98%
CAS No.: 143-07-7
Molecular Formula: CH3(CH2)10CO2H
Formula Weight: 200.32
Flash Point: 165°(329°F)
Odor: Odorless
Boiling Point: 180°/21mm
Beilstein Reference: 1099477
Merck Reference: 14,5384
MDL No.: MFCD00002736
Formula:
C₁₂H₂₄O₂ Boiling Pt: 180 °C/21 mm Melting Pt: 43…45 °C Density: 0.883 Flash Pt: 165 °C(329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002736 CAS Number: 143-07-7 EINECS: 205-582-1 Merck Index: 14,5384 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...