3-Bromo-p-toluic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11496-14
A11496-06
A11496-22
AAA11496-14EA
145
USD
AAA11496-14
AAA11496-06
AAAA11496-22
3-Bromo-p-toluic acid 98+%
3-Bromo-p-toluic acid
Formula:
C₈H₇BrO₂ Melting Pt: 206…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002488 CAS Number: 7697-26-9 EINECS: 231-712-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...