Tetrabutylphosphonium bromide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetra-n-butylphosphonium bromide
A10868-36
A10868-22
AAAA10868-36EA
210
USD
AAAA10868-36
AAAA10868-22
Tetrabutylphosphonium bromide 99%
Tetrabutylphosphonium bromide
Purity: 99%
CAS No.: 3115-68-2
Molecular Formula: C16H36BrP
Formula Weight: 339.35
Flash Point: 290°(554°F)
Melting Point: 99-103°
Beilstein Reference: 4160474
UN No.: 3464
MDL No.: MFCD00011853
Formula:
C₁₆H₃₆BrP MW: 339.34 g/mol Melting Pt: 99…103 °C Flash Pt: 290 °C (554 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011853 CAS Number: 3115-68-2 EINECS: 221-487-8 UN: 3464 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...