Iodomethane 99% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl iodide

Stabilization: Stabilised with copper

A10559-18 A10559-0B A10559-30
AAA10559-18EA 44.09 USD
AAA10559-18 AAA10559-0B AAA10559-30 AAAA10559-0B AAAA10559-30
Iodomethane 99% stabilised
Iodomethane

MDL: MFCD00001073 Beilstein Registry No.: 969135 Fieser: 1,682 2,274 3,202 4,341 5,447 6,384 8,342 9,308 19,215


Purity: 99% CAS No.: 74-88-4 Molecular Formula: CH3I Formula Weight: 141.94 Storage Temperature: 2°C to 8°C Refractive Index: 1.5300 Odor: Pungent Melting Point: -67° Boiling Point: 41-43° Beilstein Reference: 969135 Merck Reference: 14,6087 UN No.: 2644 MDL No.: MFCD00001073

Formula: CH₃I
Boiling Pt: 41…43 °C
Melting Pt: –67 °C
Density: 2.28
Storage Temperature: Refrigerator
MDL Number: MFCD00001073
CAS Number: 74-88-4
EINECS: 200-819-5
UN: 2644
ADR: 6.1,I
Merck Index: 14,6087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR