Indol-3-acetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid
A10556-14
A10556-06
A10556-22
A10556-36
AAA10556-14EA
121.33
USD
AAA10556-14
AAAA10556-06
AAA10556-22
AAA10556-36
Indol-3-acetic acid 98+%
Indol-3-acetic acid
Plant growth hormone
Purity: 98+%
CAS No.: 87-51-4
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Storage Temperature: 2°C to 8°C
Flash Point: 171°(339°F)
Melting Point: ca 167° dec.
Beilstein Reference: 143358
Merck Reference: 14,4964
MDL No.: MFCD00005636
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Boiling Pt: 120…125 °C (0.05 mmHg) Melting Pt: ∼ 167 °C (dec.) Flash Pt: 171 °C (339 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005636 CAS Number: 87-51-4 EINECS: 201-748-2 Merck Index: 14,4964 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...