Caffeine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine

A10431-22 A10431-30 A10431-0B
AAAA10431-22EA 57.53 USD
AAAA10431-22 AAA10431-30 AAA10431-0B
Caffeine 99%
Caffeine

Has inhibitory action against lung and colon tumorigenses, and virus induction


Purity: 99% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Solubility: Soluble in pyrrole, THF-H2O mixture, ethyl acetate Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758


Has inhibitory action against lung and colon tumorigenses, and virus induction

Formula: C₈H₁₀N₄O₂
MW: 194.19 g/mol
Melting Pt: 234…236 °C
Density: 1.230
Storage Temperature: Ambient
MDL Number: MFCD00005758
CAS Number: 58-08-2
EINECS: 200-362-1
UN: 1544
ADR: 6.1 III
Merck Index: 14,1636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e609e1c7a199176b9e5b938ef241d166d502f9&vid=1ac1145bba3574e2401439da90f37fee7fd7610b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR