Methyl chloroacetate 99+%

Supplier: Thermo Scientific Chemicals
Danger

A10325-0E A10325-30
AAA10325-0EEA 124 USD
AAA10325-0E AAAA10325-30
Methyl chloroacetate 99+%
Methyl chloroacetate

MDL: MFCD00000931 Beilstein Registry No.: 506255


Purity: 99+% CAS No.: 96-34-4 Molecular Formula: ClCH2CO2CH3 Formula Weight: 108.52 Flash Point: 51°(123°F) Refractive Index: 1.4220 Melting Point: -32° Boiling Point: 128-130° Beilstein Reference: 506255 Merck Reference: 14,6042 UN No.: 2295 MDL No.: MFCD00000931

Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Boiling Pt: 131
Melting Pt: -33
Density: 1.236
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000931
CAS Number: 96-34-4
EINECS: 202-501-1
UN: 2295
ADR: 6.1 I
Merck Index: 14,6042

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR