Sodium perborate tetrahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger

A10189-0E A10189-36 A10189-22
AAAA10189-0EEA 82.98 USD
AAAA10189-0E AAAA10189-36 AAAA10189-22
Sodium perborate tetrahydrate 97%
Sodium perborate tetrahydrate
Formula: NaH₂BO₄·4H₂O
Melting Pt: 60 °C (dec.)
Density: 1.73
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9
UN: 3377
ADR: 5.1,III
Merck Index: 14,8652

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR