Tricyclohexylphosphine, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

044854-03 044854-06
AAA44854-03EA 26.8 USD
AAA44854-03 AAA44854-06
Tricyclohexylphosphine, Technical Grade
Tricyclohexylphosphine

CAS No.: 2622-14-2 Molecular Formula: (C6H11)3P Formula Weight: 280.42 Physical Form: Solid Sensitivity: Air Sensitive Appearance: White to off-white Odor: Unpleasant Melting Point: 76-78° MDL No.: MFCD00003853

Formula: C₁₈H₃₃P
Boiling Pt: 132…134 °C (0.02 mmHg)
Melting Pt: 76…78 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003853
CAS Number: 2622-14-2
EINECS: 220-069-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR