Mercury(II) bromide ACS

Supplier: Thermo Scientific Chemicals
Danger

044816-22 044816-14
AAA44816-22EA 129.91 USD
AAA44816-22 AAA44816-14
Mercury(II) bromide ACS
Mercury (II) bromide

MDL: MFCD00011040 Soluble in hot alcohol, methanol, HCl2 HBr


CAS No.: 7789-47-1 Molecular Formula: Br2Hg Formula Weight: 360.40 Physical Form: Powder Density: 6.05 Sensitivity: Light Sensitive Appearance: White Odor: Odorless Melting Point: 236° Boiling Point: 322-325° subl. Solubility: Soluble in hot alcohol, methanol, HCl, HBr Merck Reference: 14,5875 UN No.: 1634 MDL No.: MFCD00011040

Formula: Br₂Hg
Boiling Pt: 322…325 °C (subl.)
Melting Pt: 236 °C
Density: 6.05
Storage Temperature: Ambient
MDL Number: MFCD00011040
CAS Number: 7789-47-1
EINECS: 232-169-3
UN: 1634
ADR: 6.1,II
Merck Index: 14,5875

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR