Soda lime, with indicator
Supplier: Thermo Scientific Chemicals
|
Danger
|
044786-36
044786-A4
AAA44786-36EA
202.13
USD
AAA44786-36
AAA44786-A4
Soda lime, with indicator
Soda lime
CAS No.: 8006-28-8
Physical Form: 8-12 Mesh
Sensitivity: Air Sensitive and Hygroscopic
Odor: Odorless
Merck Reference: 14,8569
UN No.: 1907
MDL No.: MFCD00134094
Storage Temperature:
Ambient |
MDL Number:
MFCD00134094 CAS Number: 8006-28-8 UN: 1907 ADR: 8 III Merck Index: 14,8569 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...