Aluminium oxide, γ-phase 99+%, nanopowder

Supplier: Thermo Scientific Chemicals

044757-36 044757-22 044757-14
AAA44757-36EA 662 USD
AAA44757-36 AAA44757-22 AAA44757-14
Aluminium oxide, γ-phase 99+%, nanopowder
Aluminium oxide, γ-phase

Purity: 99+% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 20nm APS Powder, S.A. 100-200m2/g Appearance: White Odor: Odorless Merck Reference: 14,356 MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR