Copper(II) oxide, nanopowder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cupric oxide, Copper oxide, Copper monoxide granulated
044663-18
044663-09
AAA44663-18EA
146
USD
AAA44663-18
AAA44663-09
Copper(II) oxide, nanopowder
Copper (II) oxide
CAS No.: 1317-38-0
Molecular Formula: CuO
Formula Weight: 79.55
Physical Form: 30-50nm APS Powder, S.A. 13m2/g
Density: 6.3
Refractive Index: 2.63
Odor: Odorless
Melting Point: 1326°
Merck Reference: 14,2646
UN No.: 3077
MDL No.: MFCD00010979
Formula:
CuO MW: 79.55 g/mol Boiling Pt: 2000 Melting Pt: 1326 Density: 6.3 Storage Temperature: Ambient |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9 III Merck Index: 14,2646 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...