Diammonium wolframate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Ammonium tungstate
044588-30
044588-09
044588-18
AAA44588-30EA
1920
USD
AAA44588-30
AAA44588-09
AAA44588-18
Diammonium wolframate ≥99.99% (metals basis)
di-Ammonium wolframate
Powder
Purity: 99.99+%
CAS No.: 11140-77-5
Molecular Formula: (NH4)2WO4
Formula Weight: 283.91
Physical Form: Powder
Solubility: Soluble in warm water
MDL No.: MFCD00015956
Formula:
H₈N₂O₄W MW: 283.91 Storage Temperature: Ambient |
MDL Number:
MFCD00015956 CAS Number: 11140-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...