Titanium diboride ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Titanium boride
044558-09
044558-30
044558-18
AAA44558-09EA
53.3
USD
AAA44558-09
AAA44558-30
AAA44558-18
Titanium diboride ≥99.5% (metals basis)
Titanium diboride
-325 Mesh Powder
Purity: 99.5%
CAS No.: 12045-63-5
Molecular Formula: B2Ti
Formula Weight: 69.50
Physical Form: -325 Mesh Powder
Density: 4.38
Appearance: Gray
Odor: Odorless
Melting Point: 3225°
MDL No.: MFCD00049595
Formula:
B₂Ti Melting Pt: 3225 °C Density: 4,38 Storage Temperature: Ambient |
MDL Number:
MFCD00049595 CAS Number: 12045-63-5 EINECS: 234-961-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...