Amidosulfonic acid ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Aminosulphonic acid, Amidosulphuric acid, Sulfamic acid, Sulfamidic acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid, Aminosulfonic acid, Amidosulfonic acid, Amidosulfuric Acid, Amidosulfuric acid
044510-14
044510-22
044510-36
AAA44510-14EA
29.58
USD
AAA44510-14
AAA44510-22
AAA44510-36
Amidosulfonic acid ≥99.99% (metals basis)
Amidosulphonic acid
Purity: 99.99%
CAS No.: 5329-14-6
Molecular Formula: H2NSO3H
Formula Weight: 97.09
Physical Form: Crystalline
Density: 2.12
pH: 1.2
Melting Point: ca 205° dec.
Merck Reference: 14,8921
UN No.: 2967
MDL No.: MFCD00011603
Formula:
H₃NO₃S MW: 97.09 g/mol Boiling Pt: 247 Melting Pt: 205 (dec.) Density: 2.12 Storage Temperature: Ambient |
MDL Number:
MFCD00011603 CAS Number: 5329-14-6 EINECS: 226-218-8 UN: 2967 ADR: 8 III Merck Index: 14,8921 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...