Platinum 10% on alumina, reduced
Supplier: Thermo Scientific Chemicals
044494-22
044494-14
AAA44494-22EA
1692
USD
AAA44494-22
AAA44494-14
Platinum 10% on alumina, reduced
Platinum
Granular
CAS No.: 7440-06-4
Physical Form: Granular
MDL No.: MFCD00011179
Formula:
Pt Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...