Lithium hydroxide, anhydrous ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

044471-06 044471-14 044471-22
AAA44471-06EA 57.6 USD
AAA44471-06 AAA44471-14 AAA44471-22
Lithium hydroxide, anhydrous ≥99.995% (metals basis)
Lithium hydroxide

Purity: 99.995% CAS No.: 1310-65-2 Molecular Formula: LiOH Formula Weight: 23.95 Physical Form: Powder Density: 2.54 Appearance: White Odor: Odorless pH: 14 Melting Point: 462° Boiling Point: 924° Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00011095

Formula: LiOH
Boiling Pt: 924 °C
Melting Pt: 462 °C
Density: 2.54
Storage Temperature: Ambient
MDL Number: MFCD00011095
CAS Number: 1310-65-2
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 14,5534

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR