Aluminium chloride, anhydrous ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044470-22
044470-14
AAA44470-22EA
508
USD
AAA44470-22
AAA44470-14
Aluminium chloride, anhydrous ≥99.999% (metals basis)
Aluminium chloride
Purity: 99.999%
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Physical Form: -10 Mesh
Density: 2.44
pH: 2,4
Melting Point: 190° subl
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422
Formula:
AlCl₃ Boiling Pt: 180 °C (1013 hPa) Melting Pt: 190 °C (subl.) Density: 2.44 Storage Temperature: Ambient |
MDL Number:
MFCD00003422 CAS Number: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8,II Merck Index: 14,337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...