2-Chlorotrityl chloride 1.0 - 1.4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh

Supplier: Thermo Scientific Chemicals

044407-09 044407-18 044407-04
AAA44407-09EA 336 USD
AAA44407-09 AAA44407-18 AAA44407-04
2-Chlorotrityl chloride 1.0 - 1.4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh
2-Chlorotrityl chloride

Powder


CAS No.: 42074-68-0 Physical Form: Powder MDL No.: MFCD00040399

Formula: C₁₉H₁₄Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00040399
CAS Number: 42074-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR