Aluminium oxide, low surface area catalyst support

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

043862-30 043862-A1 043862-A7
AAA43862-30EA 64.8 USD
AAA43862-30 AAA43862-A1 AAA43862-A7
Aluminium oxide, low surface area catalyst support
Aluminium oxide

Physical Form: 1/8" Pellets Odor: Odorless MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR