Biochanin A 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5,7-Dihydroxy-3-(4-methoxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-methoxyphenyl)chromen-4-one, 4'-Methoxy-5,7-dihydroxyisoflavone, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-4'-methoxyisoflavone
043853-03
043853-06
043853-MD
AAA43853-03EA
220
USD
AAA43853-03
AAA43853-06
AAA43853-MD
Biochanin A 98%
Biochanin A
Purity: 98%
CAS No.: 491-80-5
Molecular Formula: C16H12O5
Formula Weight: 284.27
Physical Form: Powder
Melting Point: 210-213°
Beilstein Reference: 278107
MDL No.: MFCD00006839
Formula:
C₁₆H₁₂O₅ Melting Pt: 210…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006839 CAS Number: 491-80-5 EINECS: 207-744-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...