Aluminium oxide, α-phase, low surface area catalyst support trimodal

Supplier: Thermo Scientific Chemicals

043833-A1
AAA43833-A1EA 352 USD
AAA43833-A1
Aluminium oxide, α-phase, low surface area catalyst support trimodal
Aluminium oxide, α-phase

Forme 3/16" spheres
S.A. 0.82m2/g, median pore diameter 1, 10 and 250 micron, total pore volume 0.54cc/g, packing density 0.74kg/L


Physical Form: 3/16" spheres MDL No.: MFCD00003424

Formula: Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR