Sodium hydroxide 25% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

043771-K7 043771-K2
AAA43771-K7EA 110.82 USD
AAA43771-K7 AAA43771-K2
Sodium hydroxide 25% (w/v) in aqueous solution
Sodium hydroxide

CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid UN No.: 1824 MDL No.: MFCD00003548

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR