Deuterium oxide (99.95% D) (Isotopic)
Supplier: Thermo Scientific Chemicals
Purity: 99.95+%
CAS No.: 7789-20-0
Molecular Formula: D2O
Formula Weight: 20.03
Physical Form: Liquid
Density: 1.105
Refractive Index: 1.338
Sensitivity: Moisture Sensitive
Odor: Odorless
Melting Point: 3.82°
Boiling Point: 101.4°
Merck Reference: 14,2940
MDL No.: MFCD00044636
Formula:
D₂O Boiling Pt: 101.4 °C Melting Pt: 3.82 °C Density: 1.105 Storage Temperature: Ambient |
MDL Number:
MFCD00044636 CAS Number: 7789-20-0 EINECS: 232-148-9 Merck Index: 14,2940 |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...