Zinc oxide ≥99.9% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Magnetron sputtering source
Purity: 99.9%
CAS No.: 1314-13-2
Molecular Formula: OZn
Formula Weight: 81.37
Physical Form: Target
Density: 5.606
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 1975°
UN No.: 3077
MDL No.: MFCD00011300
Magnetron sputtering source
Formula:
ZnO Melting Pt: 1975 °C Density: 5.606 Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9,III REACH: 01-2119463881-32 Merck Index: 13,10200 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...