Zinc selenide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) selenide

A19947-09 A19947-18 A19947-30
AAA19947-09EA 51.4 USD
AAA19947-09 AAA19947-18 AAA19947-30
Zinc selenide
Zinc selenide

CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Odor: Odorless Melting Point: 1100° Merck Reference: 14,10156 MDL No.: MFCD00011296

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793
Density: 5.42^1^5
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 14,10156

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR