N,N-Dimethylglycine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

A19670-14 A19670-22 A19670-06
AAA19670-14EA 285 USD
AAA19670-14 AAA19670-22 AAA19670-06
N,N-Dimethylglycine 98+%
N,N-Dimethylglycine

Purity: 98+% CAS No.: 1118-68-9 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Odor: Odorless pH: 6-7 Melting Point: 179-183° Merck Reference: 14,3244 MDL No.: MFCD00004283

Formula: C₄H₉NO₂
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8
Merck Index: 14,3244

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR