Pentafluoropyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine
A19656-06
A19656-14
AAA19656-06EA
56
USD
AAA19656-06
AAA19656-14
Pentafluoropyridine 99%
Pentafluoropyridine
Purity: 99%
CAS No.: 700-16-3
Molecular Formula: C5F5N
Formula Weight: 169.05
Flash Point: 23°(73°F)
Refractive Index: 1.3870
Boiling Point: 83-85°
Beilstein Reference: 1427064
UN No.: 2924
MDL No.: MFCD00006225
Formula:
C₅F₅N Boiling Pt: 83…85 °C Melting Pt: –41 °C Density: 1.618 Flash Pt: 23 °C (73 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006225 CAS Number: 700-16-3 EINECS: 211-839-9 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...