Ethyl phenylacetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19356-0E
A19356-22
A19356-36
AAA19356-0EEA
228
USD
AAA19356-0E
AAA19356-22
AAA19356-36
Ethyl phenylacetate 99%
Ethyl phenylacetate
Formula:
C₁₀H₁₂O₂ Boiling Pt: 223…225 °C Melting Pt: –29 °C Density: 1.030 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009178 CAS Number: 101-97-3 EINECS: 202-993-8 Merck Index: 14,3840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...