Anthrone 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Anthracen-9(10H)-one
Applications: In organic synthesis, in colorimetric determination of sugar in body fluids and of animal starch in liver tissue
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water
Purity: 97%
CAS No.: 90-44-8
Molecular Formula: C14H10O
Formula Weight: 194.23
Melting Point: 153-160°
Solubility: Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water
Beilstein Reference: 1910173
Merck Reference: 14,691
MDL No.: MFCD00001187
In organic synthesis, in colorimetric determination of sugar in body fluids and of animal starch in liver tissue
Formula:
C₁₄H₁₀O MW: 194.23 g/mol Melting Pt: 153…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001187 CAS Number: 90-44-8 EINECS: 201-994-0 Merck Index: 14,691 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...