Phosphonitrilic chloride trimer 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene

A18986-14 A18986-22
AAA18986-14EA 97.5 USD
AAA18986-14 AAA18986-22
Phosphonitrilic chloride trimer 98%
Phosphonitrilic chloride trimer

Purity: 98% CAS No.: 940-71-6 Molecular Formula: Cl6N3P3 Formula Weight: 347.66 Melting Point: 111-115° Boiling Point: 127°/13mm UN No.: 3260 MDL No.: MFCD00006474

Formula: Cl₆N₃P₃
Boiling Pt: 127 °C/13 mm
Melting Pt: 111…115 °C
Density: 1.98
Storage Temperature: Ambient
MDL Number: MFCD00006474
CAS Number: 940-71-6
EINECS: 213-376-8
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR