Ethyl methoxyacetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methoxyacetic ethyl ester
A18770-18
AAA18770-18EA
180
USD
AAA18770-18
Ethyl methoxyacetate 98%
Ethyl methoxyacetate
Purity: 98%
CAS No.: 3938-96-3
Molecular Formula: CH3OCH2CO2CH2CH3
Formula Weight: 118.13
Flash Point: 46°(115°F)
Refractive Index: 1.401
Boiling Point: 44-46°/9mm
Beilstein Reference: 1744761
UN No.: 3272
MDL No.: MFCD00009176
Formula:
C₅H₁₀O₃ MW: 118.13 g/mol Boiling Pt: 44 Density: 1.00 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009176 CAS Number: 3938-96-3 EINECS: 223-521-7 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...